Learn More
2-Methylbutyryl chloride, 99%
Description
2-Methylbutyryl chloride was used in the synthesis of 2-methylbutyramide, N-((R,S)-2-methylbutanoyl)salicylhydrazide, 2-methyl-1-phenyl-1-butanone, 1,1’-(1,3,7,9-tetrahydroxydibenzofuran-2,6-diyl)bis(2-methylbutan-1-one).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methylbutyryl chloride was used in the synthesis of 2-methylbutyramide, N-((R,S)-2-methylbutanoyl)salicylhydrazide, 2-methyl-1-phenyl-1-butanone, 1,1’-(1,3,7,9-tetrahydroxydibenzofuran-2,6-diyl)bis(2-methylbutan-1-one).
Solubility
Reacts with water.
Notes
Store away from oxidizing agents, amines, bases, alcohols, active metals, water/ moisture.
Specifications
Specifications
| Density | 0.98 |
| Boiling Point | 116°C to 117°C |
| Flash Point | 20°C (68°F) |
| Refractive Index | 1.418 |
| Quantity | 5 g |
| UN Number | UN2924 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Formula Weight | 120.58 |
| Percent Purity | 99% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.