missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutyric acid, 98%

Product Code. 11464573
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
25mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464573 2500 mL 2500mL
11349579 25 mL 25mL
11444573 100 mL 100mL
11454573 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11464573 Supplier Thermo Scientific Alfa Aesar Supplier No. A11546.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 116-53-0 | C5H10O2 | 102.133 g/mol

2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.

Solubility
It is soluble in water, but highly soluble in most common organic solvents such as Chloroform, Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Bases, Oxidizing agents, Reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-53-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00002669
InChI Key WLAMNBDJUVNPJU-UHFFFAOYSA-N
Synonym 2-methylbutyric acid, dl-2-methylbutyric acid, butanoic acid, 2-methyl, ethylmethylacetic acid, methylbutyric acid, 2-methyl butyric acid, 2-methybutyric acid, methylethylacetic acid, active valeric acid, butyric acid, 2-methyl
PubChem CID 8314
ChEBI CHEBI:37070
IUPAC Name 2-methylbutanoic acid
SMILES CCC(C)C(=O)O

Specifications

Density 0.937
Boiling Point 175°C to 176°C
Flash Point 73°C (163°F)
Assay Percent Range 98%
Refractive Index 1.4055
Quantity 2500 mL
UN Number UN3265
Beilstein 1720486
Solubility Information It is soluble in water,but highly soluble in most common organic solvents such as Chloroform,Methanol.
Formula Weight 102.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylbutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.