missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, 99+%, extra pure

Product Code. 10511754 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10511754 2.5 L 2.5 L
10542331 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10511754 Supplier Thermo Scientific Acros Supplier No. 126470025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 78-78-4 | C5H12 | 72.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -160.0°C
Color Colorless
Density 0.6200g/mL
Boiling Point 30.0°C
Flash Point −51°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(CH3)2
Refractive Index 1.3530 to 1.3550
Quantity 2.5 L
Beilstein 01, 134
Residue after Evaporation 10ppm max.
Specific Gravity 0.62
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol
Formula Weight 72.15
Vapor Pressure 990mbar at 20°C
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Methylbutane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylbutane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H224-Extremely flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.