missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl alcohol, 98%

Product Code. 11490603
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490603 50 g 50g
11480603 10 g 10g
11400613 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490603 Supplier Thermo Scientific Alfa Aesar Supplier No. A12696.18
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-95-2 | C8H10O | 122.167 g/mol

2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

Solubility
Soluble in methanol 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-95-2
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004622
InChI Key XPNGNIFUDRPBFJ-UHFFFAOYSA-N
Synonym 2-methylbenzyl alcohol, o-tolylmethanol, o-tolyl alcohol, 2-methylphenyl methanol, o-methylbenzyl alcohol, benzenemethanol, 2-methyl, toluene-methanol, benzyl alcohol, o-methyl, unii-7l3m6y04nc, alpha-hydroxy-o-xylene
PubChem CID 6994
ChEBI CHEBI:27724
IUPAC Name (2-methylphenyl)methanol
SMILES CC1=CC=CC=C1CO

Specifications

Melting Point 33°C to 37°C
Boiling Point 219°C
Flash Point 104°C (219°F)
Refractive Index 1.5408
Quantity 50 g
Beilstein 1929783
Solubility Information Soluble in methanol 0.1g/mL.
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2-Methylbenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.