missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzoxazole, 97%

Product Code. 11414624
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414624 500 g 500g
11494614 25 g 25g
11404624 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414624 Supplier Thermo Scientific Alfa Aesar Supplier No. A13198.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-21-6 | C8H7NO | 133.15 g/mol

2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

Solubility
Not miscible in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-21-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005771
InChI Key DQSHFKPKFISSNM-UHFFFAOYSA-N
Synonym 2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti
PubChem CID 7225
ChEBI CHEBI:51603
IUPAC Name 2-methyl-1,3-benzoxazole
SMILES CC1=NC2=CC=CC=C2O1

Specifications

Melting Point 2°C to 4°C
Boiling Point 200°C to 201°C
Flash Point 85°C (185°F)
Quantity 500 g
Beilstein 112297
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 2-Methylbenzoxazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.