missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazole, TRC

Product Code. 30138108
Change view
Click to view available options
Quantity:
500 mg
5 g
10 g
Unit Size:
10g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138108 10 g 10g
17937634 5 g 5g
17998784 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138108 Supplier Toronto Research Chemicals Supplier No. TRCM90000510G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-75-2
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
Synonym 2-Methylbenzothiazole, Benzothiazole, 2-methyl-, 2-Methylbenzothiazole, 2-Methyl-1,3-benzothiazole, 2-Methylbenzo[d]thiazole, NSC 3825
IUPAC Name 2-methyl-1,3-benzothiazole
SMILES Cc1nc2ccccc2s1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H7NS/c1-6-9-7-4-2-3-5-8(7)10-6/h2-5H,1H3
Formula Weight 149.0299
Chemical Name or Material 2-Methylbenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.