missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazole, 99%

Product Code. 10031840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031840 100 g Glass bottle 100g
15294634 25 g Glass bottle 25g
15204644 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10031840 Supplier Thermo Scientific Acros Supplier No. 150381000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-75-2 | C8H7NS | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-75-2
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
InChI Key DXYYSGDWQCSKKO-UHFFFAOYSA-N
Synonym 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w
PubChem CID 8446
IUPAC Name 2-methyl-1,3-benzothiazole
SMILES CC1=NC2=CC=CC=C2S1

Specifications

CAS Min % 98.5
CAS Max % 99.0
Melting Point 12.0°C to 14.0°C
Color Amber to Colorless
Density 1.2000g/mL
Boiling Point 238.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6150 to 1.6180
Quantity 100 g
Beilstein 27,46
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: ssluble in ether and chloroform
Formula Weight 149.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzothiazole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.