missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzophenone, 98+%

Product Code. 11482497
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482497 250 g 250g
11462497 10 g 10g
11472497 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482497 Supplier Thermo Scientific Alfa Aesar Supplier No. A14779.30
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 131-58-8 | C14H12O | 196.249 g/mol

2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressure.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-58-8
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008518
InChI Key CKGKXGQVRVAKEA-UHFFFAOYSA-N
Synonym 2-methylbenzophenone, o-methylbenzophenone, methanone, 2-methylphenyl phenyl, phenyl o-tolyl ketone, 2-methylphenyl phenyl methanone, benzophenone, 2-methyl, 2-methylphenyl-phenylmethanone, unii-lsn1480nff, phenyl tolyl ketone, phenyl 2-tolyl ketone
PubChem CID 67230
IUPAC Name (2-methylphenyl)-phenylmethanone
SMILES CC1=CC=CC=C1C(=O)C2=CC=CC=C2

Specifications

Melting Point -18°C
Density 1.085
Boiling Point 309°C to 311°C
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 250 g
Beilstein 2045469
Merck Index 14,7317
Solubility Information Insoluble in water.
Formula Weight 196.25
Percent Purity ≥98%
Chemical Name or Material 2-Methylbenzophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.