missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzofuran, 96%

Product Code. 11672729 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11672729 5 g Glass bottle 5g
11956201 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11672729 Supplier Thermo Scientific Acros Supplier No. 208400050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4265-25-2 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4265-25-2
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00005850
InChI Key GBGPVUAOTCNZPT-UHFFFAOYSA-N
Synonym 2-methylbenzofuran, benzofuran, 2-methyl, 2-methylcumarone, 2-methylbenzo b furan, 2-methyl benzofuran, unii-8x3183bz3x, methylbenzofuran, methyl benzoxole, 2-methyl-benzofuran, #
PubChem CID 20263
IUPAC Name 2-methyl-1-benzofuran
SMILES CC1=CC2=CC=CC=C2O1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0570g/mL
Boiling Point 197.0°C to 198.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5618
Quantity 5 g
Beilstein 17,6
Specific Gravity 1.057
Formula Weight 132.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzofuran
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylbenzofuran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.