missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzhydrol, TRC

Product Code. 30135938
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135938 10 g 10g
30012803 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135938 Supplier Toronto Research Chemicals Supplier No. TRCM28892010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5472-13-9
Molecular Formula C14 H14 O
Molecular Weight (g/mol) 198.26
Synonym (RS)-(2-Methylphenyl)phenylmethanol, Benzenemethanol, 2-methyl-α-phenyl-, Benzhydrol, 2-methyl- (6CI,7CI,8CI), 2-Methyl-α-phenylbenzenemethanol, (2-Methylphenyl)phenylmethanol, 2-Methylbenzhydrol, NSC 27910, Phenyl(2-tolyl)methanol, Phenyl(o-tolyl)methanol, o-Methylbenzhydrol, α-(2-Methylphenyl)benzenemethanol, α-(2-Methylphenyl)benzyl alcohol
IUPAC Name (2-methylphenyl)-phenylmethanol
SMILES Cc1ccccc1C(O)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H14O/c1-11-7-5-6-10-13(11)14(15)12-8-3-2-4-9-12/h2-10,14-15H,1H3
Formula Weight 198.1045
Chemical Name or Material (RS)-(2-Methylphenyl)phenylmethanol (2-Methylbenzhydrol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.