missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzhydrol, 98+%, Thermo Scientific™

Product Code. 11476196
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476196 25 g 25g
11486196 100 g 100g
11496196 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476196 Supplier Thermo Scientific Alfa Aesar Supplier No. B23021.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5472-13-9
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00004490
InChI Key MXHXXJOHFRHBFB-UHFFFAOYSA-N
Synonym 2-methylbenzhydrol, phenyl o-tolyl methanol, benzhydrol, 2-methyl, 2-methylphenyl phenylmethanol, o-methylbenzhydrol, phenyl-o-tolyl-methanol, 2-methylphenyl-phenylmethanol, benzenemethanol, 2-methyl-.alpha.-phenyl, 2-methylphenyl phenyl methanol, phenyl-o-tolylcarbinol
PubChem CID 95659
IUPAC Name (2-methylphenyl)-phenylmethanol
SMILES CC1=CC=CC=C1C(C2=CC=CC=C2)O

Specifikationer

Melting Point 93°C to 95°C
Boiling Point 323°C
Quantity 25 g
Beilstein 1952703
Formula Weight 198.27
Percent Purity ≥98%
Chemical Name or Material 2-Methylbenzhydrol
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.