missing translation for 'onlineSavingsMsg'
Learn More

2-Methylanisole, TRC

Product Code. 30482699
Change view
Click to view available options
Quantity:
150 g
Unit Size:
150g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482699 150 g 150g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482699 Supplier Toronto Research Chemicals Supplier No. TRCM266330150G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methylanisole
CAS 578-58-5
Molecular Formula C8H10O
Quantity 150 g
Synonym o-Methylanisole, 1-Methoxy-2-methylbenzene, 2-Cresol Methyl Ether, 2-Methoxytoluene, 2-Methyl-1-methoxybenzene, Methyl o-Cresyl Ether, Methyl o-Methylphenyl ether, Methyl o-Tolyl Ether, NSC 6253, o-Cresol Methyl Ether, o-Cresyl Methyl Ether, o-Methoxytoluene, o-Methylanisole
InChI Formula InChI=1S/C8H10O/c1-7-5-3-4-6-8(7)9-2/h3-6H,1-2H3
SMILES COc1ccccc1C
IUPAC Name 1-methoxy-2-methylbenzene
Molecular Weight (g/mol) 122.16
Formula Weight 122.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.