missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester, 96%

Product Code. 15471476
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471476 250 mg 250mg
15461476 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471476 Supplier Thermo Scientific Alfa Aesar Supplier No. H54682.MD
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 904326-88-1 | C11H18BN3O2 | 235.094 g/mol

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 904326-88-1
Molecular Formula C11H18BN3O2
Molecular Weight (g/mol) 235.094
MDL Number MFCD11878345
InChI Key QDOXNCAIXITTKA-UHFFFAOYSA-N
Synonym n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid, pinacol ester, n-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid,pinacol ester, 2-methylamino pyrimidine-5-boronic acid pinacol ester, 2-methylamino pyrimidin-5-yl boronic acid pinacol ester, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl-amine, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl amine, n-m ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, n-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-amine
PubChem CID 46739641
IUPAC Name N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC

Specifications

Melting Point 122°C to 125°C
Quantity 250 mg
Formula Weight 235.09
Percent Purity 96%
Chemical Name or Material 2-(Methylamino)pyrimidine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.