missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)ethanol, TRC

Product Code. 30137733
Change view
Click to view available options
Quantity:
5 mL
50 mL
100 mL
Unit Size:
100mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137733 100 mL 100mL
30137394 50 mL 50mL
30003448 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137733 Supplier Toronto Research Chemicals Supplier No. TRCM286090100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-83-1
Molecular Formula C3 H9 N O
Molecular Weight (g/mol) 75.1097
Synonym 2-(Methylamino)ethanol (ACI), (2-Hydroxyethyl)methylamine, (Hydroxyethyl)methylamine, 2-(N-Methylamino)ethanol, 2-Hydroxy-N-methylethylamine, 2-Hydroxyethyl-N-methylamine, 2-Methylamino-1-ethanol, 2-N-Monomethylaminoethanol, Amietol M 11, Amino Alcohol MMA, Methyl(2-hydroxyethyl)amine, Methyl(hydroxyethyl)amine, Methyl(β-hydroxyethyl)amine, Methylaminoethanol, Methylethanolamine, Methylethylolamine, Monomethylaminoethanol, Monomethylethanolamine, Monomethylmonoethanolamine, N-(2-Hydroxyethyl)-N-methylamine, N-(2-Hydroxyethyl)methylamine, N-Methyl-2-aminoethanol, N-Methyl-2-ethanolamine, N-Methyl-2-hydroxyethanamine, N-Methyl-2-hydroxyethylamine, N-Methyl-N-(2-hydroxyethyl)amine, N-Methyl-N-(β-hydroxyethyl)amine, N-Methyl-N-hydroxyethylamine, N-Methylaminoethanol, N-Methylethanolamine, N-Methylmonoethanolamine, N-Monomethylaminoethanol, N-Monomethylethanolamine, NMEA, NSC 62776, Yubao, β-(Methylamino)ethanol, 2-(Methylamino)ethanol
IUPAC Name 2-(methylamino)ethanol
SMILES CNCCO

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C3H9NO/c1-4-2-3-5/h4-5H,2-3H2,1H3
Formula Weight 75.0684
Percent Purity >95%
Chemical Name or Material N-methylethanolamine
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.