missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)ethanol, 99%

Product Code. 10351474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351474 10 kg Metal Drum 10kg
10522751 500 g Glass bottle 500g
10367773 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351474 Supplier Thermo Scientific Acros Supplier No. 158260100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-83-1 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-83-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00002839
InChI Key OPKOKAMJFNKNAS-UHFFFAOYSA-N
Synonym 2-methylamino ethanol, n-methylethanolamine, methylethanolamine, ethanol, 2-methylamino, methylethylolamine, n-methylaminoethanol, n-methylmonoethanolamine, n-monomethylethanolamine, 2-hydroxyethyl methylamine, monomethylaminoethanol
PubChem CID 8016
ChEBI CHEBI:21763
IUPAC Name 2-(methylamino)ethanol
SMILES CNCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -3.0°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 159.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Metal Drum
Linear Formula CH3NHCH2CH2OH
Refractive Index 1.4380 to 1.4400
Quantity 10 kg
Beilstein 04, 276
Merck Index 15, 6089
Specific Gravity 0.93
Formula Weight 75.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Methylamino)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.