missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)ethanol, 99%

Product Code. 11420777
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420777 100 mL 100mL
11430777 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420777 Supplier Thermo Scientific Alfa Aesar Supplier No. B23180.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109-83-1 | C3H9NO | 75.11 g/mol

2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.

Solubility
Miscible with water, alcohol and ether.

Notes
Incompatible with oxidizing agents, acids, copper, copper alloys, zinc and galvanized iron.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-83-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00002839
InChI Key OPKOKAMJFNKNAS-UHFFFAOYSA-N
Synonym 2-methylamino ethanol, n-methylethanolamine, methylethanolamine, ethanol, 2-methylamino, methylethylolamine, n-methylaminoethanol, n-methylmonoethanolamine, n-monomethylethanolamine, 2-hydroxyethyl methylamine, monomethylaminoethanol
PubChem CID 8016
ChEBI CHEBI:21763
IUPAC Name 2-(methylamino)ethanol
SMILES CNCCO

Specifications

Melting Point -5°C
Density 0.939
Boiling Point 159°C
Flash Point 74°C (165°F)
Odor Ammonia-like
Linear Formula CH3NH(CH2)2OH
Refractive Index 1.44
Quantity 100 mL
UN Number UN2735
Beilstein 1071196
Merck Index 14,6015
Solubility Information Miscible with water,alcohol and ether.
Formula Weight 75.11
Percent Purity 99%
Chemical Name or Material 2-(Methylamino)ethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-(Methylamino)ethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: N-Methylethanolamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.