missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)-1H-purin-6(7H)-one, MedChemExpress

Product Code. 16403358
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16403358

Brand: Medchem Express HY101412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids.

2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids.

Specifications

Chemical Name or Material 2-(Methylamino)-1H-purin-6(7H)-one
Physical Form Solid
CAS 10030-78-1
Color Earth Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C6H7N5O
Synonym N2-methylguanine
Solubility Information H2O : 1 mg/mL (6.06 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (6.06 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble) ∣THF : < 1 mg/mL (ultrasonic) (insoluble) ∣1M NaOH : < 1 mg/mL (ultrasonic;adjust pH to 12 with NaOH) (insoluble) ∣Acetone : < 1 mg/mL (ultrasonic) (insoluble) ∣0.1 M HCL : < 1 mg/mL (ultrasonic;adjust pH to 2 with HCl) (insoluble) ∣DMF : < 1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1N=C(NC)NC2=C1NC=N2
Molecular Weight (g/mol) 165.15
Formula Weight 165.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.