missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetophenone, 98%

Product Code. 11379579
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379579 5 g 5g
11459203 25 g 25g
11469203 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379579 Supplier Thermo Scientific Alfa Aesar Supplier No. A11695.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 577-16-2 | C9H10O | 134.178 g/mol

2'-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-16-2
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00008734
InChI Key YXWWHNCQZBVZPV-UHFFFAOYSA-N
Synonym 2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
PubChem CID 11340
IUPAC Name 1-(2-methylphenyl)ethanone
SMILES CC1=CC=CC=C1C(=O)C

Specifications

Density 1.02
Boiling Point 214°C
Flash Point 75°C (167°F)
Refractive Index 1.5318
Quantity 5 g
Beilstein 907005
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 2'-Methylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.