missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetophenone, 98%

Product Code. 10606522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10606522 25 g Glass bottle 25g
10722515 5 g Glass bottle 5g
10147953 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10606522 Supplier Thermo Scientific Acros Supplier No. 126160250
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 577-16-2 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-16-2
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008734
InChI Key YXWWHNCQZBVZPV-UHFFFAOYSA-N
Synonym 2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
PubChem CID 11340
IUPAC Name 1-(2-methylphenyl)ethanone
SMILES CC1=CC=CC=C1C(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0260g/mL
Boiling Point 214°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4COCH3
Refractive Index 1.5290 to 1.533
Quantity 25 g
Beilstein 07, 306
Specific Gravity 1.026
Formula Weight 134.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2'-Methylacetophenone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.