missing translation for 'onlineSavingsMsg'
Learn More

2-[Methyl(phenylmethyl)amino]ethanol, TRC

Product Code. 30138519
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138519 10 g 10g
30136934 25 g 25g
30005888 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138519 Supplier Toronto Research Chemicals Supplier No. TRCM32046510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-98-4
Molecular Formula C10 H15 N O
Molecular Weight (g/mol) 165.23
Synonym Ethanol, 2-[methyl(phenylmethyl)amino]-, Ethanol, 2-(benzylmethylamino)- (6CI,7CI,8CI), 2-[Methyl(phenylmethyl)amino]ethanol, 2-(Benzylmethylamino)ethanol, 2-(N-Methylbenzylamino)ethanol, 2-(N-benzyl-N-methylamino)ethanol, Benzyl(2-hydroxyethyl)methylamine, N-Benzyl-N-methyl(2-hydroxyethyl)amine, N-Benzyl-N-methyl-2-aminoethanol, N-Benzyl-N-methylethanolamine, N-Methyl-N-benzylethanolamine, β-(N-Benzyl-N-methylamino)ethanol
IUPAC Name 2-[benzyl(methyl)amino]ethanol
SMILES CN(CCO)Cc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H15NO/c1-11(7-8-12)9-10-5-3-2-4-6-10/h2-6,12H,7-9H2,1H3
Formula Weight 165.1154
Chemical Name or Material N-Benzyl-N-methyl-ethanolamine (2-[benzyl(methyl)amino]ethanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.