missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals

Product Code. 15483686
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
This item is not returnable. View return policy

Product Code. 15483686

Brand: Thermo Scientific Alfa Aesar H57776.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (1.00E+06 mg/L).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1158098-73-7
Molecular Formula C7H13NO2S
Molecular Weight (g/mol) 175.246
MDL Number MFCD14702522
InChI Key VKUZMNXQGKBLHN-UHFFFAOYSA-N
Synonym 2-methyl-n-oxetan-3-ylidene propane-2-sulfinamide, 2-methyl-n-3-oxetanylidene-2-propanesulfinamide, 2-methyl-n-3-oxetanylidene propane-2-sulfinamide, 3-tert-butylsulfinylmethylene oxetane, n-tert-butylsulfinyl oxetane-3-imine, 2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide, 2-methyl-n-oxetan-3-ylidene ;propane-2-sulfinamide, 2-methyl-propane-2-sulfinic acid oxetan-3-ylideneamide, 2-methylpropane-2-sulfinic acid oxetan-3-ylideneamide
PubChem CID 46839995
IUPAC Name 2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
SMILES CC(C)(C)S(=O)N=C1COC1

Specifications

Quantity 250 mg
Solubility Information Soluble in water. (1.00E+06mg/L).
Formula Weight 175.25
Percent Purity 95%
Chemical Name or Material 2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.