missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Methyl-DL-serine hydrate, 96%

Product Code. p-168547 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10123631

Brand: Thermo Scientific Chemicals 339750010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5424-29-3 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5424-29-3
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD03791288
InChI Key CDUUKBXTEOFITR-UHFFFAOYNA-N
Synonym 2-methylserine, alpha-methyl-dl-serine, dl-2-methylserine, alpha-methylserine, 2-methyl-dl-serine, dl-serine, 2-methyl, dl-alpha-methylserine, dl-.alpha.-methylserine, dl-2-methylserine hydrate, .alpha.-methyl-dl-serine
PubChem CID 94309
ChEBI CHEBI:17799
IUPAC Name 2-amino-3-hydroxy-2-methylpropanoic acid
SMILES CC(N)(CO)C(O)=O

Specifications

Melting Point 253.0°C to 263.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids
Formula Weight 119.12
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-DL-serine hydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.