missing translation for 'onlineSavingsMsg'
Learn More

(2-methyl-benzoimidazol-1-yl)-acetic acid, 97%, Thermo Scientific™

Product Code. 10061054
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10061054 10 g 10g
10001014 250 mg 250mg
10537284 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10061054 Supplier Thermo Scientific Maybridge Supplier No. MO08104EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40332-17-0
Molecular Formula C10H9N2O2
Molecular Weight (g/mol) 189.20
MDL Number MFCD00621731
InChI Key SMGBDFGKVGSCGB-UHFFFAOYSA-M
Synonym 2-methyl-1h-benzimidazol-1-yl acetic acid, 2-2-methyl-1h-benzo d imidazol-1-yl acetic acid, 2-methyl-benzoimidazol-1-yl-acetic acid, 2-methyl-1h-benzimidazole-1-acetic acid, 1h-benzimidazole-1-acetic acid, 2-methyl, 2-methylbenzoimidazol-1-yl acetic acid, 2-methyl-benzoimidazol-1-yl acetic acid, 2-2-methylbenzimidazol-1-yl acetic acid, 2-methyl-1,3-benzodiazol-1-yl acetic acid, 2-2-methylbenzimidazolyl acetic acid
PubChem CID 719681
SMILES CC1=NC2=CC=CC=C2N1CC([O-])=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name 2-(2-methyl-1H-1,3-benzodiazol-1-yl)acetate
Formula Weight 190.2
Percent Purity 97%
Chemical Name or Material (2-Methyl-benzoimidazol-1-yl)-acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.