missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-9H-fluoren-9-one, TRC

Codice prodotto. 30482884
Cambia vista
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Dimensione della confezione:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30482884 500 mg 500mg
30471788 100 mg 100mg
30472865 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30482884 Fornitore Toronto Research Chemicals N. del fornitore TRCM321793500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2840-51-9
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
IUPAC Name 2-methylfluoren-9-one
SMILES Cc1ccc2c3ccccc3C(=O)c2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H10O/c1-9-6-7-11-10-4-2-3-5-12(10)14(15)13(11)8-9/h2-8H,1H3
Formula Weight 194.0732
Chemical Name or Material 2-Methyl-9H-fluoren-9-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.