missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 15498315
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498315 1 g 1g
15408325 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498315 Supplier Thermo Scientific Alfa Aesar Supplier No. H33110.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 545394-83-0 | Molecular Formula: C8H6F3NO | Molecular Weight: 189.14 g/mol

Participate in Horner-Wadsworth-Emmons reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Participate in Horner-Wadsworth-Emmons reaction.

Solubility
Sparingly Soluble (6.1E-4 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545394-83-0
Molecular Formula C8H6F3NO
Molecular Weight (g/mol) 189.14
MDL Number MFCD11035943
InChI Key HHGVNSFXEFHMGQ-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl nicotinaldehyde, 2-methyl-6-trifluoromethyl pyridine-3-carbaldehyde, 2-methyl-6-trifluoromethyl pyridine-3-carboxaldehyde, 2-methyl-6-trifluoromethyl-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 2-methyl-6-trifluoromethyl
PubChem CID 28875322
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde
SMILES CC1=C(C=O)C=CC(=N1)C(F)(F)F

Specifications

Melting Point 45°C to 48°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble (6.1E-4g/L) (25°C).
Formula Weight 189.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.