missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)nicotinic acid, Tech., Thermo Scientific™

Product Code. 10659604
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10659604 1 g Amber Glass Bottle 1g
10230133 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10659604 Supplier Thermo Scientific Maybridge Supplier No. AW00680DA
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 261635-93-2
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD01311990
InChI Key ODWJFUQRQUCRJZ-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl nicotinic acid, 2-methyl-6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethyl-2-methylpyridine-3-carboxylic acid, 2-methyl-6-trifluoromethyl-nicotinic acid, maybridge4_000139, 2-methyl-6-trifluoromethynicotinic acid, 2-methyl-6-trifluoromethyinicotinic acid, 2-methyl-6-trifluoromethylnicotinic acid, 2-methyl-6-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 2775683
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES CC1=C(C=CC(=N1)C(F)(F)F)C(O)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.13
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.