missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)nicotinic acid, 97%

Product Code. 15455787
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455787 1 g 1g
15465787 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455787 Supplier Thermo Scientific Alfa Aesar Supplier No. H33003.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 261635-93-2 | C8H6F3NO2 | 205.14 g/mol

2-Methyl-6-(trifluoromethyl)nicotinic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-6-(trifluoromethyl)nicotinic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water (4.7 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261635-93-2
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD01311990
InChI Key ODWJFUQRQUCRJZ-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl nicotinic acid, 2-methyl-6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethyl-2-methylpyridine-3-carboxylic acid, 2-methyl-6-trifluoromethyl-nicotinic acid, maybridge4_000139, 2-methyl-6-trifluoromethynicotinic acid, 2-methyl-6-trifluoromethyinicotinic acid, 2-methyl-6-trifluoromethylnicotinic acid, 2-methyl-6-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 2775683
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES CC1=C(C=CC(=N1)C(F)(F)F)C(O)=O

Specifications

Melting Point 139°C to 141°C
Quantity 1 g
Solubility Information Slightly soluble in water (4.7g/L at 25°C).
Formula Weight 205.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)nicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.