missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)nicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10763512
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10763512 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10763512 Supplier Thermo Scientific Maybridge Supplier No. MAY00323DA
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 545394-83-0
Assay Percent Range 97%
Molecular Formula C8H6F3NO
MDL Number MFCD11035943
Quantity 1 g
Synonym 2-methyl-6-trifluoromethyl nicotinaldehyde, 2-methyl-6-trifluoromethyl pyridine-3-carbaldehyde, 2-methyl-6-trifluoromethyl pyridine-3-carboxaldehyde, 2-methyl-6-trifluoromethyl-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 2-methyl-6-trifluoromethyl
InChI Key HHGVNSFXEFHMGQ-UHFFFAOYSA-N
SMILES CC1=C(C=O)C=CC(=N1)C(F)(F)F
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde
Molecular Weight (g/mol) 189.14
PubChem CID 28875322
Formula Weight 189.14
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)nicotinaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.