missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-6-quinolinyl)methanol, 97%, Thermo Scientific™

Catalog Number 10015902
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
108166-02-5
C11H11NO
173.22
MFCD06659066
HLFYJILNIBADGG-UHFFFAOYSA-N
2-methylquinolin-6-yl methanol, 2-methyl-6-quinolinyl methanol, 2-methylquinoline-6-methanol, 6-quinolinemethanol,2-methyl, 6-quinolinemethanol, 2-methyl, 6-hydroxymethyl-2-methylquinoline, acmc-1bsb5, 2-methyl-6-quinolyl methan-1-ol, 6-hydroxymethyl-2-methyl-1-azanaphthalene
2795481
(2-methylquinolin-6-yl)methanol
CC1=CC=C2C=C(CO)C=CC2=N1
This item is not returnable. View return policy
108166-02-5
C11H11NO
173.22
MFCD06659066
HLFYJILNIBADGG-UHFFFAOYSA-N
2-methylquinolin-6-yl methanol, 2-methyl-6-quinolinyl methanol, 2-methylquinoline-6-methanol, 6-quinolinemethanol,2-methyl, 6-quinolinemethanol, 2-methyl, 6-hydroxymethyl-2-methylquinoline, acmc-1bsb5, 2-methyl-6-quinolyl methan-1-ol, 6-hydroxymethyl-2-methyl-1-azanaphthalene
2795481
(2-methylquinolin-6-yl)methanol
CC1=CC=C2C=C(CO)C=CC2=N1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material (2-Methyl-6-quinolinyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'