missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitrobenzoic acid, 99%, Thermo Scientific™

Product Code. 15565046 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15565046 25 g 25g
10143750 1 g 1g
10153750 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15565046 Supplier Thermo Scientific Acros Supplier No. 165980250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13506-76-8
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
InChI Key CCXSGQZMYLXTOI-UHFFFAOYSA-N
Synonym 2-methyl-6-nitrobenzoic acid, 6-nitro-o-toluic acid, benzoic acid, 2-methyl-6-nitro, 6-methyl-2-nitrobenzoic acid, 2-methyl-6-nitro-benzoic acid, o-toluic acid, 6-nitro, 2-methyl-6-nitrobenzoicacid, unii-jm85y56dx1, nitrotoluylsaure, nitro-ortho-toluic acid
PubChem CID 16097
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 154.0°C to 157.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CH3C6H3(NO2)CO2H
Quantity 25 g
Beilstein 09,I,189
Solubility Information Solubility in water: <0.1g/100mL (20°C).
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 2-Methyl-6-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.