missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-methoxypyridine-3-boronic acid, 97%

Product Code. 11915891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11915891 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11915891 Supplier Thermo Scientific Acros Supplier No. 431420010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 459856-12-3 | C7H10BNO3 | 166.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 459856-12-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point 231.2°C to 232.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H10BNO3
MDL Number MFCD05662386
Quantity 1 g
Synonym 2-methyl-6-methoxypyridine-3-boronic acid, 6-methoxy-2-methylpyridin-3-yl boronic acid, 6-methoxy-2-picoline-3-boronic acid, 6-methoxy-2-methylpyridine-3-boronic acid, 6-methoxy-2-methylpyridin-3-yl boranediol, 3-borono-6-methoxy-2-methylpyridine, boronic acid, 6-methoxy-2-methyl-3-pyridinyl, 2-methoxy-6-methyl-5-pyridinylboronic acid, 6-methoxy-2-methyl-3-pyridineboronic acid, boronic acid,b-6-methoxy-2-methyl-3-pyridinyl
InChI Key CWXWIICBKIHZFC-UHFFFAOYSA-N
SMILES COC1=NC(C)=C(C=C1)B(O)O
IUPAC Name (6-methoxy-2-methylpyridin-3-yl)boronic acid
Molecular Weight (g/mol) 166.97
PubChem CID 2763084
Formula Weight 166.97
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Methyl-6-methoxypyridine-3-boronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-6-methoxypyridine-3-boronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.