missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenyl-3-furyl isocyanate, Thermo Scientific™

Product Code. 10225762
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225762 1 g Amber Glass Bottle 1g
10767804 250 mg Amber Glass Bottle 250mg
10577844 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10225762 Supplier Thermo Scientific Maybridge Supplier No. CC14906DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 568577-82-2
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
InChI Key LVNIEPBRJLDJHU-UHFFFAOYSA-N
Synonym 2-methyl-5-phenyl-3-furyl isocyanate, 2-methyl-5-phenyl-3-furylisocyanate, 2-methyl-5-phenylfuran-3-isocyanate, 3-isocyanato-2-methyl-5-phenyl-furan
PubChem CID 2795315
IUPAC Name 3-isocyanato-2-methyl-5-phenylfuran
SMILES CC1=C(C=C(O1)C2=CC=CC=C2)N=C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 2-Methyl-5-phenyl-3-furyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.