missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenyl-3-furoic acid, 97%, Thermo Scientific™

Product Code. 10512852
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10512852 1 g Amber Glass Bottle 1g
10522852 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10512852 Supplier Thermo Scientific Maybridge Supplier No. CC14901DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 108124-17-0
Molecular Formula C12H9O3
Molecular Weight (g/mol) 201.20
MDL Number MFCD00221070
InChI Key VLMNACSEESRUAK-UHFFFAOYSA-M
Synonym 2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid
PubChem CID 736516
IUPAC Name 2-methyl-5-phenylfuran-3-carboxylic acid
SMILES CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-phenyl-3-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.