missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitropyridine, 95%, Thermo Scientific Chemicals

Catalog Number 15428318
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
21203-68-9
C6H6N2O2
138.126
MFCD04114179
USZINSZJSVMICC-UHFFFAOYSA-N
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036
2794552
2-methyl-5-nitropyridine
CC1=NC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
21203-68-9
C6H6N2O2
138.126
MFCD04114179
USZINSZJSVMICC-UHFFFAOYSA-N
5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036
2794552
2-methyl-5-nitropyridine
CC1=NC=C(C=C1)[N+](=O)[O-]

2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 138.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Methyl-5-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'