missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrophenylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. p-4380099 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11996111

Brand: Thermo Scientific Acros 445170010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 957062-84-9
Molecular Formula C13H18BNO4
Molecular Weight (g/mol) 263.1
InChI Key LYFUUPOOXAXEEP-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-2-methyl-5-nitrophenyl-1,3,2-dioxaborolane, 2-methyl-5-nitrophenylboronic acid pinacol ester, 2-methyl-5-nitrophenylboronic acid, pinacol ester, 2-methyl-5-nitrophenyl boric acid pinacol ester, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 2-methyl-5-nitrophenylboronate, pubchem18757, 2-methyl-5-nitrophenylboronicacidpinacolester, 2-methyl-5-nitrobenzeneboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2-methyl-5-nitro-phenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-methyl-5-nitrophenyl
PubChem CID 17750269
IUPAC Name 4,4,5,5-tetramethyl-2-(2-methyl-5-nitrophenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)[N+](=O)[O-])C

Specifications

Melting Point 90.0°C to 92.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 263.1
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-nitrophenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Methyl-5-nitrophenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.