missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-5-nitroimidazole, 99%

Product Code. 15447098
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447098 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447098 Supplier Alfa Aesar™ Supplier No. H60614.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
2-Methyl-4(5)-nitroimidazole was used to study the mechanism of both the alkaline and acidic hydrolysis of tinidazole. It is mainly used to synthesize intermediate of medicines such as Metronidazole and dimetridazole.

Solubility
Slightly soluble in ethanol, easily soluble in diluted acid and diluted alkali.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88054-22-2
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00005191
InChI Key FFYTTYVSDVWNMY-UHFFFAOYSA-N
Synonym 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
PubChem CID 12760
IUPAC Name 2-methyl-5-nitro-1H-imidazole
SMILES CC1=NC=C(N1)[N+]([O-])=O

Specifications

Melting Point 251°C to 255°C
Density 1.426g/mL
Boiling Point 399°C
Quantity 250 g
Beilstein 4032
Formula Weight 127.1
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 2-Methyl-5-nitroimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.