missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzoic Acid, TRC

Product Code. 30464576
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464576 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464576 Supplier Toronto Research Chemicals Supplier No. TRCM32368510G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methyl-5-nitrobenzoic Acid
CAS 1975-52-6
Molecular Formula C8 H7 N O4
Quantity 10 g
Synonym Benzoic acid','2-methyl-5-nitro-, o-Toluic acid','5-nitro- (6CI,7CI,8CI), 2-Methyl-5-nitrobenzoic acid, 3-Nitro-6-methylbenzoic acid, 5-Nitro-2-methylbenzoic acid, 5-Nitro-o-toluic acid
InChI Formula InChI=1S/C8H7NO4/c1-5-2-3-6(9(12)13)4-7(5)8(10)11/h2-4H,1H3,(H,10,11)
SMILES Cc1ccc(cc1C(=O)O)[N+](=O)[O-]
IUPAC Name 2-methyl-5-nitrobenzoic acid
Molecular Weight (g/mol) 181.15
Formula Weight 181.0375
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.