missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzoic acid, 99+%, Thermo Scientific™

Product Code. 10105553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10105553 25 g Glass bottle 25g
10007352 1 g Glass bottle 1g
10227220 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10105553 Supplier Thermo Scientific Acros Supplier No. 276570250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1975-52-6
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007371
InChI Key DJRFJAVPROZZFL-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, 2-methyl-5-nitro-benzoic acid, 5-nitro-2-methylbenzoic acid, rarechem al bo 0285, 5-nitro-2-methyl benzoic acid, 2-methyl-5-nitro benzoic acid, pubchem2244
PubChem CID 519683
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 176°C to 180°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)CO2H
Quantity 25 g
Beilstein 09, 471
Solubility Information Solubility in water: insoluble
Formula Weight 181.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitrobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.