missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzoic acid, 99+%, ACROS Organics™

Brand:  Acros Organics 276570050

Code : 29

Additional Details : CAS Number : 1975-52-6 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10227220

  • £43.60 / 5g


Estimated Shipment
18-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-5-nitrobenzoic acid
99.0
≥99.0%
C8H7NO4
181.15
5-Nitro-o-toluic acid
1975-52-6
100.0
99+%
CH3C6H3(NO2)CO2H
MFCD00007371

Specifications

2-Methyl-5-nitrobenzoic acid
99.0
Authentic
Crystalline Powder
99+%
CH3C6H3(NO2)CO2H
MFCD00007371
Yellow to beige
176° to 180°C
1975-52-6
100.0
Glass bottle
≥99.0%
C8H7NO4
181.15
09, 471
5-Nitro-o-toluic acid
5g
Videos
SDS