missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzenesulfonyl chloride, 97%

Product Code. 11457816
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457816 25 g 25g
11447816 5 g 5g
11467816 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457816 Supplier Thermo Scientific Alfa Aesar Supplier No. A14760.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-02-8 | C7H6ClNO4S | 235.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-02-8
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.64
MDL Number MFCD00051695
InChI Key WPGVQDHXOUAJBW-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzene-1-sulfonyl chloride, 5-nitro-o-toluenesulfonyl chloride, 2-methyl-5-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methyl-5-nitro, 4-nitrotoluen-2-sulfochlorid, o-toluenesulfonyl chloride, 5-nitro, 4-nitrotoluen-2-sulfonylchlorid, 4-nitrotoluene-2-sulphonyl chloride, 4-nitrotoluen-2-sulfochlorid czech, 2-methyl-5-nitrophenylsulfonyl chloride
PubChem CID 8457
IUPAC Name 2-methyl-5-nitrobenzenesulfonyl chloride
SMILES CC1=CC=C(C=C1S(Cl)(=O)=O)[N+]([O-])=O

Specifications

Melting Point 42°C to 46°C
Boiling Point 135°C to 137°C (0.7 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 2697079
Sensitivity Moisture sensitive
Formula Weight 235.65
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-nitrobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.