missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroaniline, TRC

Product Code. 30138444
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138444 10 g 10g
30133592 50 g 50g
30040393 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138444 Supplier Toronto Research Chemicals Supplier No. TRCM32344510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-55-8
Molecular Formula C7 H8 N2 O2
Molecular Weight (g/mol) 152.15
Synonym Fast Scarlet G Base (6CI), o-Toluidine, 5-nitro- (8CI), 2-Methyl-5-nitrobenzenamine, (2-Methyl-5-nitrophenyl)amine, 1-Amino-2-methyl-5-nitrobenzene, 1-Methyl-2-amino-4-nitrobenzene, 2-Amino-4-nitrotoluene, 2-Methyl-5-nitroaniline, 3-Nitro-6-methylaniline, 4-Nitro-2-aminotoluene, 5-Nitro-2-methylaniline, 5-Nitro-2-toluidine, 5-Nitro-o-toluidine, 6-Methyl-3-nitroaniline, Amarthol Fast Scarlet G Base, Amarthol Fast Scarlet G Salt, Azoene Fast Scarlet G Salt, Azoene Fast Scarlet GC Base, Azoene Fast Scarlet GC Salt, Azofix Scarlet G Salt, Azogene Fast Scarlet G, C.I. 37105, C.I. Azoic Diazo Component 12, Conazoic Diazo AB, Dainichi Fast Scarlet G Base, Daito Scarlet Base G, Devol Scarlet B, Devol Scarlet G Salt, Diabase Scarlet G, Diazo Fast Scarlet G, Dycosbase Scarlet G Base, Fast Red SG Base, Fast Scarlet Base J, Fast Scarlet G, Fast Scarlet G Salt, Fast Scarlet GC Base, Fast Scarlet J Salt, Fast Scarlet M 4NT Base, Fast Scarlet T Base, Hiltonil Fast Scarlet G Base, Hiltonil Fast Scarlet G Salt, Hiltonil Fast Scarlet GC Base, Icho Salt Scarlet G, Kayaku Scarlet G Base, Lake Scarlet G Base, Lithosol Orange R Base, Mitsui Scarlet G Base, NSC 8947, Naphthanil Scarlet G Base, Naphtoelan Fast Scarlet G Base, Naphtoelan Fast Scarlet G Salt, PNOT, Scarlet Base Ciba II, Scarlet Base G, Scarlet Base Irga II, Scarlet Base NSP, Scarlet G, Scarlet G Base, Sugai Fast Scarlet G Base, Symulon Scarlet G Base
IUPAC Name 2-methyl-5-nitroaniline
SMILES Cc1ccc(cc1N)[N+](=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H8N2O2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,8H2,1H3
Formula Weight 152.0586
Chemical Name or Material 2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.