missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroaniline, 98+%

Product Code. 11463318
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11463318

Brand: Thermo Scientific Alfa Aesar A17726.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-55-8 | C7H8N2O2 | 152.15 g/mol

It is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.

Solubility
Insoluble in water. (<0.1 g/100 mL at 19°C). Solubility in methanol, almost transparency

Notes
C.I. 37105. Sensitive to moisture and light. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-55-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007741
InChI Key DSBIJCMXAIKKKI-UHFFFAOYSA-N
Synonym 2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
PubChem CID 7444
ChEBI CHEBI:66891
IUPAC Name 2-methyl-5-nitroaniline
SMILES CC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

Melting Point 105°C to 108°C
Odor Characteristic
Quantity 500 g
UN Number UN2660
Beilstein 879021
Solubility Information Insoluble in water. (<0.1g/100 mL at 19°C). Solubility in methanol,almost transparency
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2-Methyl-5-nitroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.