missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitro-N-[4-(pyridin-3-yl)pyrimidin-2-yl]benzenamine, Mikromol

Product Code. 30460344
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30460344

Brand: Mikromol MM3566.0400254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Methyl-5-nitro-N-[4-(pyridin-3-yl)pyrimidin-2-yl]benzenamine, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidine-amine
CAS 152460-09-8
Analyte or Component Names N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidine-amine
Shipping Condition Room Temperature
Molecular Formula C16 H13 N5 O2
Certifications/Compliance ISO 9001
Synonym 2-Pyrimidinamine, N-(2-methyl-5-nitrophenyl)-4-(3-pyridinyl)-, (2-Methyl-5-nitrophenyl)[4-(pyridin-3-yl)pyrimidin-2-yl]amine, 2-methyl-5-nitro-N-[4-(pyridin-3-yl)pyrimidin-2-yl]benzenamine, N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidineamine, N-(2-Methyl-5-nitrophenyl)-4-(pyridin-3-yl)pyrimidin-2-amine, N-(5-Nitro-2-methylphenyl)-N-[4-(3-pyridyl)pyrimidin-2-yl]amine, N-(5-Nitro-2-methylphenyl)[4-(3-pyridyl)-2-pyrimidinyl]amine
InChI Formula InChI=1S/C16H13N5O2/c1-11-4-5-13(21(22)23)9-15(11)20-16-18-8-6-14(19-16)12-3-2-7-17-10-12/h2-10H,1H3,(H,18,19,20)
Solution Type Neat
SMILES Cc1ccc(cc1Nc2nccc(n2)c3cccnc3)[N+](=O)[O-]
IUPAC Name N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
Molecular Weight (g/mol) 307.31
Quantity 4 x 25 mg
Formula Weight 307.1069 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.