missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-5-hydroxytryptamine hydrochloride, Tocris Bioscience™
5-HT3 agonist/potent 5-HT6 ligand
Chemical Identifiers
CAS | 845861-49-6 |
---|---|
Molecular Formula | C11H15ClN2O |
Molecular Weight (g/mol) | 226.704 |
InChI Key | ZNVRHQPXSBWLQP-UHFFFAOYSA-N |
Synonym | 2-methylserotonin.hcl, 2-methyl-5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-2-methyl-1h-indol-5-ol hydrochloride, 2-methyl-5-ht hydrochloride |
PubChem CID | 11492320 |
IUPAC Name | 3-(2-aminoethyl)-2-methyl-1H-indol-5-ol;hydrochloride |
SMILES | CC1=C(C2=C(N1)C=CC(=C2)O)CCN.Cl |
Description
5-HT3 agonist (Ki = 1200 nM) and potent 5-HT6 ligand (Ki = 46 nM).Chemical Identifiers
845861-49-6 | |
226.704 | |
2-methylserotonin.hcl, 2-methyl-5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-2-methyl-1h-indol-5-ol hydrochloride, 2-methyl-5-ht hydrochloride | |
3-(2-aminoethyl)-2-methyl-1H-indol-5-ol;hydrochloride |
C11H15ClN2O | |
ZNVRHQPXSBWLQP-UHFFFAOYSA-N | |
11492320 | |
CC1=C(C2=C(N1)C=CC(=C2)O)CCN.Cl |
Specifications
845861-49-6 | |
2-methylserotonin.hcl, 2-methyl-5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-2-methyl-1h-indol-5-ol hydrochloride, 2-methyl-5-ht hydrochloride | |
CC1=C(C2=C(N1)C=CC(=C2)O)CCN.Cl | |
226.704 | |
226.71 | |
2-Methyl-5-hydroxytryptamine hydrochloride |
C11H15ClN2O | |
ZNVRHQPXSBWLQP-UHFFFAOYSA-N | |
3-(2-aminoethyl)-2-methyl-1H-indol-5-ol;hydrochloride | |
11492320 | |
>98% |
Safety and Handling
Recommended Storage : Store at Room Temperature