missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-pentenoic acid, 98%

Product Code. 11483674
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483674 10 g 10g
11493674 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483674 Supplier Thermo Scientific Alfa Aesar Supplier No. A11297.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1575-74-2 | C6H10O2 | 114.144 g/mol

2-Methyl-4-pentenoic acid is used as raw material for fruit flavor or it is an enhancer of fruity fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-4-pentenoic acid is used as raw material for fruit flavor or it is an enhancer of fruity fragrances.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1575-74-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00015570
InChI Key HVRZYSHVZOELOH-UHFFFAOYSA-N
Synonym 2-methyl-4-pentenoic acid, 4-pentenoic acid, 2-methyl, 2-methylpent-4-en-1-oic acid, fema no. 3511, 2-methyl-4-pentenoicacid, methylallylacetic acid, acmc-209dg9, 2-methyl-pent-4-enoic acid, hvrzyshvzoeloh-uhfffaoysa, 2-methyl-4-pentenoic acid, fg
PubChem CID 549519
IUPAC Name 2-methylpent-4-enoic acid
SMILES CC(CC=C)C(=O)O

Specifications

Density 0.95
Boiling Point 195°C
Flash Point 107°C (224°F)
Odor Characteristic
Refractive Index 1.431
Quantity 10 g
UN Number UN3265
Beilstein 1720987
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-4-pentenoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.