missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-octanone, 99%

Product Code. 11468817
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468817 25 g 25g
11458817 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468817 Supplier Thermo Scientific Alfa Aesar Supplier No. A19952.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7492-38-8 | C9H18O | 142.24 g/mol

2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7492-38-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00048808
InChI Key ICSKJDZASFIJQK-UHFFFAOYSA-N
Synonym 2-methyl-4-octanone, 4-octanone, 2-methyl
PubChem CID 522566
IUPAC Name 2-methyloctan-4-one
SMILES CCCCC(=O)CC(C)C

Specifications

Boiling Point 180°C
Refractive Index 1.415
Quantity 25 g
Formula Weight 142.24
Percent Purity 99%
Chemical Name or Material 2-Methyl-4-octanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.