missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-nitroaniline, 98%

Product Code. 11428486
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428486 2500 g 2500g
11408486 100 g 100g
11418486 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428486 Supplier Thermo Scientific Alfa Aesar Supplier No. A15941.0E

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-52-5 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-52-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007734
InChI Key XTTIQGSLJBWVIV-UHFFFAOYSA-N
Synonym 2-amino-5-nitrotoluene, 4-nitro-o-toluidine, 5-nitro-2-aminotoluene, p-nitro-o-toluidine, benzenamine, 2-methyl-4-nitro, fast red rl base, 1-amino-2-methyl-4-nitrobenzene, diabase red rl, ansibases red rl, devol red rl
PubChem CID 7441
IUPAC Name 2-methyl-4-nitroaniline
SMILES CC1=CC(=CC=C1N)[N+]([O-])=O

Specifications

Melting Point 130°C to 133°C
Odor Characteristic
Quantity 2500 g
UN Number UN2660
Beilstein 775772
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-4-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.