missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-isothiazolin-3-one, 50% aqueous solution

Product Code. 15589503 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15589503

Brand: Thermo Scientific Acros 459970050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2682-20-4 | C4H5NOS | 115.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2682-20-4
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
InChI Key BEGLCMHJXHIJLR-UHFFFAOYSA-N
Synonym 2-methyl-4-isothiazolin-3-one, methylisothiazolinone, 2-methyl-4-isothiazoline-3-one, 2-methylisothiazol-3 2h-one, 2-methyl-3 2h-isothiazolone, n-methylisothiazolone, 3 2h-isothiazolone, 2-methyl, caswell no. 572a, 2-methyl-3-isothiazolone, unii-229d0e1qfa
PubChem CID 39800
ChEBI CHEBI:53620
IUPAC Name 2-methyl-1,2-thiazol-3-one
SMILES CN1C(=O)C=CS1

Specifications

Density 1.1600g/mL
pH 5.0 to 7.0 (1% aq. soln.)
Quantity 5 g
Specific Gravity 1.16
Formula Weight 115.15
Chemical Name or Material 2-Methyl-4-isothiazolin-3-one
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-4-isothiazolin-3-one, 50% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.