missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(trifluoromethyl)aniline, 98%, Thermo Scientific™

Product Code. 11410461
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410461 5 g 5g
11400461 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410461 Supplier Thermo Scientific Alfa Aesar Supplier No. B20009.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54396-44-0
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD00153216
InChI Key TWLDBACVSHADLI-UHFFFAOYSA-N
Synonym 2-methyl-3-trifluoromethyl aniline, 3-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-o-toluidine, 3-amino-2-methyl benzotrifluoride, 2-methyl-3-trifluoromethyl-phenylamine, 2-methyl-3 trifluoromethyl aniline, 2-methyl-3-trifluoromethyl benzenamine, benzenamine, 2-methyl-3-trifluoromethyl
PubChem CID 2735932
IUPAC Name 2-methyl-3-(trifluoromethyl)aniline
SMILES CC1=C(C=CC=C1N)C(F)(F)F

Specifications

Melting Point 37°C to 40°C
Boiling Point 62°C to 64°C (4 mmHg)
Flash Point 98°C (208°F)
Quantity 5 g
UN Number UN2811
Beilstein 2832865
Formula Weight 175.16
Percent Purity 98%
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.