missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-pentanol, 97+%

Product Code. 11337945
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337945 10 g 10g
11347945 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337945 Supplier Thermo Scientific Alfa Aesar Supplier No. L08015.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 565-67-3 | C6H14O | 102.177 g/mol

2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

Solubility
Amalgamate with ethanol, ethyl ether, benzene. Insoluble in water

Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-67-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004567
InChI Key ISTJMQSHILQAEC-UHFFFAOYSA-N
Synonym 2-methyl-3-pentanol, 3-pentanol, 2-methyl, propanol, 1-isopropyl, acmc-20anqw, 4-methyl-3-pentanol, 2-methyl-pentan-3-ol, 4-01-00-01716 beilstein handbook reference, istjmqshilqaec-uhfffaoysa
PubChem CID 11264
IUPAC Name 2-methylpentan-3-ol
SMILES CCC(C(C)C)O

Specifications

Density 0.83
Boiling Point 127°C to 129°C
Flash Point 46°C (114°F)
Refractive Index 1.417
Quantity 10 g
UN Number UN2282
Beilstein 1718952
Solubility Information Amalgamate with ethanol,ethyl ether,benzene. Insoluble in water
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Methyl-3-pentanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.